Wasser, ultrapure HPLC

Lieferant: Thermo Fisher Scientific

Synonyme: Wasserstoffoxid, Hydrogenoxid

22934.K2 22934.K7 22934.M6
22934.K2EA 53.3 EUR
22934.K2 22934.K7 22934.M6
Wasser, ultrapure HPLC
Wasser

CAS No.: 7732-18-5 Molecular Formula: H2O Formula Weight: 18.02 Physical Form: Liquid Density: 1.000 Refractive Index: 1.3330 Odour: Odourless Melting Point: 0° Boiling Point: 100° Merck Reference: 14,10039 MDL No.: MFCD00011332

Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100 °C
Schmelzpunkt: 0
Dichte: 1.000
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 14,10039

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Clear colorless
Form Liquid
Assay (unspecified) Specific Conductance at 25°C and time packaging: 1.5 microS/cm, max
Optical Absorbance at 400 nm 0.005 AU, max- at 300 nm 0.005 AU, max- at 250 nm 0.005 AU, max- at 200 nm 0.010 AU, max- at 190 nm 0.010 AU, max
Residue on Evaporation 2 ppm, Max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR