Magnesiumaluminat ≥99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
Synonyme:
Magnesiumaluminiumoxid, Dialuminiummagnesiumtetraoxid, Aluminiummagnesiumoxid
22950.22
22950.36
22950.PG
22950.22EA
50.1
EUR
22950.22
22950.36
22950.PG
Magnesiumaluminat ≥99% (Metall-Basis)
Magnesium aluminate
Purity: 99%
CAS No.: 12068-51-8
Molecular Formula: MgAl2O4
Formula Weight: 142.26
Physical Form: -325 Mesh Powder
Odour: Odourless
MDL No.: MFCD00049475
Formel:
Al₂MgO₄ Molecular Weight: 142.27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049475 CAS-Nummer: 12068-51-8 EINECS: 235-100-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...