Vanadium(IV)oxid ≥99% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

22957.09 22957.18
22957.09EA 93.9 EUR
22957.09 22957.18
Vanadium(IV)oxid ≥99% (Metall-Basis)
Vanadium(IV)oxid

Purity: 99% CAS No.: 12036-21-4 Molecular Formula: O2V, may contain V6O13 Formula Weight: 82.94 Physical Form: -100 Mesh Powder Density: 4.339 Appearance: Black Melting Point: 1967° UN No.: 3285 MDL No.: MFCD00049702

Formel: O₂V
Siedepunkt: 2700 °C (1013 hPa)
Schmelzpunkt: 1967 °C
Dichte: 4,339
Lagertemperatur: Raumtemperatur
MDL: MFCD00049702
CAS-Nummer: 12036-21-4
EINECS: 234-841-1
UN: 3285
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR