Acid red 52, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Food Red No.106, Sodium 4-(3,6-bis(diethylamino)xanthylium-9-yl)benzene-1,3-disulphonate, Kiton Red S, Sulphorhodamine B sodium salt, Lissamine rhodamine B-200

230162-5G
230162-5GEA 170 EUR
230162-5G
Acid red 52, Sigma-Aldrich®
Acid red 52

Sulforhodamine B has been used as a dye in the MCF-7 (human breast adenocarcinoma cell line) cell proliferation assay.

Formel: C₂₇H₃₀N₂O₇S₂
Molecular Weight: 580.65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010180
CAS-Nummer: 3520-42-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b2646cb5450239b6724d0d8f6d53e42908176ce&vid=561bbd9120a3de393a607486eaddd0a5ab5db9cf&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR