1,3-Diphenylaceton-p-toluolsulfonylhydrazon ≥98%, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
230308-10G
230308-10GEA
218
EUR
230308-10G
1,3-Diphenylaceton-p-toluolsulfonylhydrazon ≥98%, Sigma-Aldrich®
1,3-Diphenylaceton-p-toluolsulfonylhydrazon
Formel:
C₂₂H₂₂N₂O₂S Schmelzpunkt: 187,..188 °C (Lit.) Lagertemperatur: Room Temperature |
MDL:
MFCD00009645 CAS-Nummer: 19816-88-7 EINECS: 243-344-9 |
|
Spezifikation Testergebnisse
Appearance (Form) | Conforms to Requirements |
Infrared spectrum | Conforms to Structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...