Viktoriareinblau BO 90% (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Basic Blue 7, N-(4-((4-(Diethylamino)phenyl)(4-(ethylamino)naphthalen-1-yl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium chloride

M-Clarity™ Qualitätsstufe = MQ100

230987-25G
230987-25GEA 223 EUR
230987-25G
Viktoriareinblau BO 90% (Farbstoffgehalt), Sigma-Aldrich®
Viktoriareinblau BO

Victoria blue dyes are triaminodiphenylnaphthylmethanes, with lipophilic cations and large conjugated systems.

Formel: C₃₃H₄₀ClN₃
Molecular Weight: 514,1438 g/mol
Lagertemperatur: Room Temperature
MDL: MFCD00011877
CAS-Nummer: 2390-60-5
EINECS: 219-232-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf6b83e06fb78169e3c55a8092d47b67534e8cb0&vid=cf6b83e06fb78169e3c55a8092d47b67534e8cb0&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Brown to Very Dark Brown and Brown-Yellow and Yellow-Brown
Appearance (Form) Powder or Crystal or Chunk(s)
Infrared spectrum Conforms to Structure
Wavelength c = 0.008g/L; Water 612 - 618 nm
Extinction Coefficient ≥62000
Carbon 68.0 - 86.3 %
Nitrogen 7.2 - 9.1 %
Solubility (Color) 0.1 % in 95% Ethanol:Water (1:1) Dark Blue to Very Dark Blue

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR