Spatulas
Lieferant: Agencinox
1822 1826 1869 1975
231-0000EA 147 EUR
231-0000 231-0001 231-0002 231-0003
Spatulas
Spatel
Stainless steel ¹⁸/₁₀, anti-magnetic, electro-polishing. These universal spatulas are suitable for many applications.
  • Excellent chemical and thermal resistance
  • Autoclavable