Tiegelzangen
Lieferant: NICKEL ELECTRO
3021
231-0353EA 0 EUR
231-0353
Tiegelzangen
Zangen
Pure nickel (99,6%). With grooved tips. For easy, safe handling of crucibles made from a wide variety of materials.
  • Excellent corrosion resistance
  • Good thermal and electrical conductivity
  • Excellent thermal resistance