Tiegelzangen
Lieferant: NICKEL ELECTRO
3041
231-0354EA 7.9 EUR
231-0354
Tiegelzangen
Zangen
Steel, non alloy. For easy, safe handling of crucibles made from a wide variety of materials.
  • With grooved tips
  • Good thermal resistance
  • Robust and easy action