Tiegelzangen
Lieferant: NICKEL ELECTRO
3051
231-0356EA 47.2 EUR
231-0356
Tiegelzangen
Zangen
Pure nickel (99,6%). For easy, safe handling of crucibles made from a wide variety of materials.
  • With grooved tips
  • Excellent corrosion resistance
  • Good thermal and electrical conductivity
  • Excellent thermal resistance