Stainless steel ¹⁸/₁₀, anti-magnetic. Useful combination of instruments used daily in the laboratory and in production.
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Comes in practical imitation leather case
Each set includes a 130×4 mm micro double spatula, a 150×9 mm powder spatula, a pair of laboratory scissors (sharp/blunt, 130 mm long), as well as forceps (straight, sharp, 130 mm long).
Jedes Set enthält einen 130×4 mm-Mikro-Doppelspatel, einen 150×9 mm-Pulverspatel, eine Laborschere (scharf/stumpf, 130 mm Länge) sowie eine Pinzette (gerade, scharf, 130 mm Länge).