Cobalt(II)iodid, wasserfrei ≥99,5% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99.5%
CAS No.: 15238-00-3
Molecular Formula: CoI2
Formula Weight: 312.74
Physical Form: -60 Mesh Powder
Sensitivity: Light Sensitive and Hygroscopic
Boiling Point: 570° vac.
Solubility: Soluble in water
Merck Reference: 14,2443
MDL No.: MFCD00016030
Moisture and humidity indicator, catalyst
Formel:
CoI₂ Siedepunkt: 570 °C (in Vacuum) Dichte: 5,68 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016030 CAS-Nummer: 15238-00-3 EINECS: 239-283-2 Merck Index: 14,2443 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...