Cobalt(II)acetat Co ≥ 32%, wasserfrei
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
CAS No.: 71-48-7
Molecular Formula: C4H6CoO4
Formula Weight: 177.02
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Pink
Odour: Odourless
Melting Point: 298° dec.
Solubility: Soluble in water
Merck Reference: 14,2433
UN No.: 3077
MDL No.: MFCD00008689
Formel:
C₄H₆CoO₄ Dichte: 1,7755 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008689 CAS-Nummer: 71-48-7 EINECS: 200-755-8 UN: 3077 ADR: 9,III Merck Index: 14,2433 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...