Cobalt(II) tetrafluoroborate hydrate ≥96%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
23142.14
23142.22
23142.14EA
30
EUR
23142.14
23142.22
Cobalt(II) tetrafluoroborate hydrate ≥96%
Cobalt(II) tetrafluoroborate hydrate
Purity: 96%
CAS No.: 26490-63-1
Molecular Formula: Co(BF4)2.xH2O
Formula Weight: 232.54(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
UN No.: UN3260
MDL No.: MFCD00016012
Formel:
Co(BF₄)₂·xH₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016012 CAS-Nummer: 26490-63-1 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...