Entomological forceps
Lieferant: MURRAY S
E009/01
232-0038EA 11.2 EUR
232-0038
Entomological forceps
Pinzetten
Stainless steel. Forceps for the collection of entomological specimens.
  • Guide pin helps to keep the tips aligned
  • Wide tips help to avoid damages of the specimen
  • Curved tips won't crush delicate larvae or eggs