ESD Safe rubber handle tweezers
Lieferant: OUTILS RUBIS
00-SA-RH 2A-SA-RH 2B-SA-RH 3-SA-RH 3C-SA-RH 5-SA-RH 5A-SA-RH 5AR-SA-RH 7-SA-RH
232-0232EA 47.6 EUR
232-0232 232-0233 232-0234 232-0235 232-0236 232-0237 232-0238 232-0239 232-0240
ESD Safe rubber handle tweezers
Pinzetten
Streamlined tweezers with rubber handles to grant a secure grip, even after several hours of work.
  • Made from anti-magnetic stainless steel, with TPV rubber handles
  • Excellent ESD properties
  • Special streamlined shape fits the human hand perfectly
TPV rubber material has been developed with high-tech materials to provide heat insulating, effortless handling, extended durability, and ESD safe material (10⁷ Ohm). Rubies streamlined rubber handles are particularly recommended for cleanrooms and other controlled environments. They also provide good thermic insulation from heat, which makes them perfect for soldering and desoldering applications.