Forceps, ceramic
Lieferant: SLG KUNSTOFFABRIK UND FORMENBA
392507
232-1644EA 167 EUR
232-1644
Forceps, ceramic
Pinzetten
Straight ceramic forceps with serrated tips.
  • Anti-magnetic, corrosion-free, metal-free
  • Dishwasher-safe
  • Chemically resistant to aggressive media
  • Extremely heat-resistant
  • Non-wetting