Micro preparation forceps
Lieferant: HAMMACHER KARL
HWC111-10
232-1917EA 63.8 EUR
232-1917
Micro preparation forceps
Pinzetten
Wironit steel (18/12 stainless steel with 18% chrome and 10 - 12% nickel).
  • Resistant to corrosion
  • Anti-magnetic
  • Very delicate, curved