Cyclohexan ≥99,5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

23224.293EA 43.7 EUR
23224.293 23224.327 23224.362 23224.464 23224.550
Cyclohexan ≥99,5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens
Cyclohexan
Formel: C₆H₁₂
Siedepunkt: 81 °C (1013 hPa)
Schmelzpunkt: 7 °C
Dichte: 0,7785 g/cm³ (20 °C)
Flammpunkt: –18 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003814
CAS-Nummer: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
REACH: 01-2119463273-41
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (on anhydrous substance) Min. 99.5 %
IR Spectrum Passes test
Readily carbonisable substances Passes test
Boiling point 79 - 81 °C
Colouration Max. 10 APHA
Density (20/4) 0.778 - 0.779
Density (20/20) 0.779 - 0.780
Free acidity Max. 0.0003 meq/g
Solidification point 5.5 - 6.5 °C
Substances coloured by H2SO4 Max. 60 APHA
Benzene Max. 100 ppm
Cyclohexene Max. 100 ppm
Evaporation residue Max. 10 ppm
Water Max. 100 ppm
Al (Aluminium) Max. 0.1 ppm
Ba (Barium) Max. 0.02 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0,06 ppm
Na (Sodium) Max. 1 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.05 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.1 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR