Minor skin graft blade, No. SG3
Lieferant: SWANN MORTON
2201
233-0053EA 33.6 EUR
233-0053
Minor skin graft blade, No. SG3
Skalpelle Skalpellklingen
Stainless steel. Originally developed for use with skin grafts, these blades are also suitable for a wide range of other applications.
  • Fits standard No. 3 surgical handle (233-5454)
  • Sterile
  • Individually packed
Ideal for use on the hands and feet of small children given its practical size advantage compared to a standard skin grafting knife. Also suitable for use within Histology departments for the fine sectioning of the pathology blocks.
Verpackung: Packed in individual foil packs.