Trapezförmige Klinge, Griff aus massivem Zinkdruckguss.
- Full metal housing: very robust and abrasion resistant
- Interchangeable blade (tool required)
- With spring-loaded blade retraction and blade control slider on top
- Trapezoid deep-edged installed blade
- Blade with two cutting edges doubles the efficiency of the cutter
Heavy-duty cutter for use by left and right-handed users. The spring-loaded blade retract system is designed to respond to any uncontrolled move if and when the thumb-push slider is not in forward position. Ideal for cutting tape, plastic, cardboard, textile, rubber, PVC and leather.
TÜV approved
Standards: TÜV-geprüft