Multi-purpose scissors, Rubis®
Lieferant: OUTILS RUBIS
1C200/EL
233-0218EA 42.7 EUR
233-0218
Multi-purpose scissors, Rubis®
Scheren
Long legged multi-purpose scissors with rounded-off tips.
  • Produced from hardened stainless steel AISI420
  • Highly resistant against acids and alkaline
  • Ergonomically designed Rubis Twist® grips
  • No Medical Device (MD) product
Made in Switzerland to the highest quality standards. Rubis products are the result of traditional craftsmanship, strict throughput controls and a finishing process under a magnifying glass. No tool leaves the factory without having passed the strict quality controls. Neither scratches nor inaccuracies in material or surface are permitted.