Long legged precision scissors made from the best surgical steel.
Produced from hardened stainless steel AISI420
Highly resistant against acids and alkaline
With ergonomically designed Rubis Twist® grips
No Medical Device (MD) product
Made in Switzerland to the highest quality standards. Rubis products are the result of traditional craftsmanship, strict throughput controls and a finishing process under the magnifying glass. No tool leaves the factory without having passed the strict quality controls. Neither scratches nor inaccuracies in material or surface are permitted.