Scissors, duck, Rubis®
Lieferant: OUTILS RUBIS
1F003/EL
233-0224EA 32 EUR
233-0224
Scissors, duck, Rubis®
Scheren
Micro scissors with beak-shaped tips.
  • Produced from hardened stainless steel AISI420
  • Highly resistant against acids and alkaline
  • With ergonomically designed Rubis Twist® grips
  • No Medical Device (MD) product
Made in Switzerland to the highest quality standards. Rubis products are the result of traditional craftsmanship, strict throughput controls and a finishing process under the magnifying glass. No tool leaves the factory without having passed the strict quality controls. Neither scratches nor inaccuracies in material or surface are permitted.