Scissors with tweezer tips, Rubis®
Lieferant: OUTILS RUBIS
1K603
233-0226EA 31.6 EUR
233-0226
Scissors with tweezer tips, Rubis®
Scheren
Allround tweezers engineered into a scissors-movement.
  • Produced from hardened stainless steel AISI420
  • Highly resistant against acids and alkaline
  • With ergonomically designed Rubis Twist® grips and high precison tweezers tips
  • No Medical Device (MD) product
Made in Switzerland to the highest quality standards. Rubis products are the result of traditional craftsmanship, strict throughput controls and a finishing process under the magnifying glass. No tool leaves the factory without having passed the strict quality controls. Neither scratches nor inaccuracies in material or surface are permitted.