Dithiothreitol (DTT, Cleland's Reagenz) 99, Calbiochem®
Lieferant: Merck Millipore Calbiochem
|
Achtung
|
Synonyme:
DTT, (R*,R*)-1,4-Dimercapto-2,3-butandiol
233153-1
233153-10
233153-5
233153-1EA
140
EUR
233153-1
233153-10
233153-5
Dithiothreitol (DTT, Cleland's Reagenz) 99, Calbiochem®
Dithiothreitol (DTT, Cleland's Reagenz)
Formel:
C₄H₁₀O₂S₂ Molecular Weight: 154.25 g/mol Siedepunkt: ∼125 °C (1013 hPa) Schmelzpunkt: 42…44 °C Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00004877 CAS-Nummer: 3483-12-3 EINECS: 222-468-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...