1,1-Dichlor-1-fluorethan ≥99,5%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

23370.297EA 0 EUR
23370.297
1,1-Dichlor-1-fluorethan ≥99,5%, AnalaR NORMAPUR® analytisches Reagens
1,1-Dichlor-1-fluorethan
Formel: Cl₂FCCH₃
Siedepunkt: 31,9 °C (1013 hPa)
Schmelzpunkt: –103,5 °C
Dichte: 1,24 g/cm³ (20 °C)
CAS-Nummer: 1717-00-6
EINECS: 404-080-1
UN: 3077
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Assay Min. 99.5 %
Acidity Max. 0.0002 meq/g
Colouration Max. 10 APHA
Carbonyl compounds (as CH3COCH3) Max. 10 ppm
Evaporation residue Max. 5 ppm
Free chlorine Max. 1 ppm
Ionised chlorine Max. 0.5 ppm
Water Max. 50 ppm
Cd (Cadmium) Max. 0.01 ppm
Cu (Copper) Max. 0.01 ppm
Fe (Iron) Max. 0.1 ppm
Ni (Nickel) Max. 0.01 ppm
Pb (Lead) Max. 0.01 ppm
Zn (Zinc) Max. 0.01 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR