Diethanolamin 98,5-101,0%, GPR RECTAPUR®
Lieferant: VWR Chemicals
|
Gefahr
|
23372.290EA
40.9
EUR
23372.290
23372.360
23372.460
Diethanolamin 98,5-101,0%, GPR RECTAPUR®
Diethanolamin
Formel:
C₄H₁₁NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002843 CAS-Nummer: 111-42-2 EINECS: 203-868-0 REACH: 01-2119488930-28 |
Spezifikation Testergebnisse
Assay (calculated on anhydrous) | 98.5 - 101.0 % |
Identification (IR) Passes test | Passes test |
Melting point (lower value) | ≥ 25 °C |
Melting point (upper value) | ≤ 29 °C |
Water | ≤ 0.15 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...