Coelenterazine ≥90% (durch TLC), Calbiochem®, Sigma-Aldrich®

Lieferant: Merck Millipore Calbiochem


M-Clarity™ Qualitätsstufe = MQ100

233900-250
233900-250EA 430 EUR
233900-250
Coelenterazine ≥90% (durch TLC), Calbiochem®, Sigma-Aldrich®
Coelenterazine

Coelenterazine (CTZ) is a cell-permeable aequorin luminophore that acts as a very sensitive and specific chemiluminescence probe of the superoxide anion. It can also be used for detecting changes in intracellular Ca2+ in cells that have been transfected with apoaequorin cDNA.


  • Packaged under inert gas


CTZ functions as a powerful antioxidant. It serves as a substrate for multidrug resistance protein 1 (MDR1) P-glycoprotein (ATP-binding cassette sub-family B member 1 (ABCB1)/Pgp) and can bind with apoaequorin to form the active aequorin complex. It can be mainly used to quantify the formation of superoxide in neutrophils.

Formel: C₂₆H₂₁N₃O₃
Lagertemperatur: Gefrierschrank
MDL: MFCD00467176
CAS-Nummer: 55779-48-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR