Erythrosin gelblich, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung


M-Clarity™ Qualitätsstufe = MQ100

234079-25G
234079-25GEA 199 EUR
234079-25G
Erythrosin gelblich, Sigma-Aldrich®
Erythrosin gelblich
Formel: C₂₀H₈I₂Na₂O₅
Molecular Weight: 628.07 g/mol
Lagertemperatur: Room Temperature
MDL: MFCD00005045
CAS-Nummer: 33239-19-9
EINECS: 251-419-2

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Red
Appearance (Form) Conforms to Requirements
Powder or Crystals
Infrared spectrum Conforms to Structure
Wavelength 522 - 528 nm
Absorbance ≥0.85
TLC Consistent with Past Lots
Solubility (Turbidity) Clear
10 mg/ml, H₂O
Solubility (Colour) Red

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR