Indigotrisulphonic acid potassium salt ≥60% (durch HPLC) ozonsammelndes Reagens, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Potassium indigotrisulphonate

234087-1G 234087-5G
234087-1GEA 112 EUR
234087-1G 234087-5G
Indigotrisulphonic acid potassium salt ≥60% (durch HPLC) ozonsammelndes Reagens, Sigma-Aldrich®
Indigotrisulphonic acid potassium salt

Ozone-scavenging reagent that reacts rapidly with ozone. Potassium indigotrisulfonate has been used: In colorimetric method of measurement of atmospheric ozone, for measuring residual aqueous ozone by gas diffusion flow injection analysis, to determine O₃ concentration in water.

Formel: C₁₆H₇K₃N₂O₁₁S₃
Molecular Weight: 616.71 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013160
CAS-Nummer: 67627-18-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=091776922f775454785cbc5d758781fad9a25367&vid=1c3c2ba6684ff1ceaf647a19f5e142aae8a5d733&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Dark Blue to Very Dark Blue and Dark Purple to Very Dark Purple and Black
Appearance (Form) Powder or Crystals or Powder with Chunk(s) or Crystals with Chunk(s)
Infrared spectrum Conforms to Structure
Wavelength 250 - 256 nm
Extinction Coefficient ≥ 23000
Extinction Coefficient ( c = 0.01g/l in H2O) ≥ 17000
Purity (HPLC) ≥ 60 %
Carbon Typically 19.5% - 26.0%
Nitrogen Typically 2.5% - 4.0%
Solubility (Color) c = 0.1%; Water Dark Blue to Very Dark Blue


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR