Oil Red EGN, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

234117-25G
234117-25GEA 93.2 EUR
234117-25G
Oil Red EGN, Sigma-Aldrich®
Oil Red EGN

Oil Red EGN has been used to stain lipid droplets accumulated in cell during adipogenesis.

Formel: C₂₅H₂₂N₄O
Schmelzpunkt: 130 °C (Zers., Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009688
CAS-Nummer: 4477-79-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d09f39eb43c54453ca858fdf983b8beff1d301b0&vid=e78333e5f50c6f7d6b0e0c74950f5354e5b27a77&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR