Dimethylammoniumchlorid, TECHNICAL
Lieferant: VWR Chemicals
|
Achtung
|
23431.232EA
0
EUR
23431.232
Dimethylammoniumchlorid, TECHNICAL
Dimethylammoniumchlorid
Formel:
C₂H₈ClN Schmelzpunkt: 170…173 °C |
MDL:
MFCD00012477 CAS-Nummer: 506-59-2 EINECS: 208-046-5 Merck Index: 13,03255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...