Kongorot, Sigma-Aldrich®
Lieferant: Merck Millipore Calbiochem
|
Gefahr
|
234610-500
234610-500EA
126
EUR
234610-500
Kongorot, Sigma-Aldrich®
Kongorot
Formel:
C₃₂H₂₂N₆Na₂O₆S₂ Schmelzpunkt: > 360 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004028 CAS-Nummer: 573-58-0 EINECS: 209-358-4 Merck Index: 12,02562 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...