N,N-Dimethylformamid ≥99,8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

23466.298EA 40.4 EUR
23466.298 23466.323 23466.550 23466.460 23466.930
N,N-Dimethylformamid ≥99,8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytisches Reagens
N,N-Dimethylformamid
Formel: HCON(CH₃)₂
Siedepunkt: 153 °C (1013 hPa)
Schmelzpunkt: –60,5 °C
Dichte: 0,949 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003284
CAS-Nummer: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (on anhydrous substance) Min. 99.8 %
IR Spectrum Passes test
Acidity Max. 0.00025 meq/g
Alkalinity Max. 0.003 meq/g
Boiling point 152 - 155 °C
Colouration Max. 10 APHA
Density (20/4) 0.946 - 0.950
Density (20/20) 0.949 - 0.952
Evaporation residue Max. 10 ppm
Water Max. 0.05 %
Al (Aluminium) Max. 0.5 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium)
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 0.5 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.1 ppm
Sn (Tin) Max. 0.01 ppm
Sr (Strontium) Max. 0.05 ppm
Zn (Zinc)
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR