N,N-Dimethylformamid ≥99,5%, GPR RECTAPUR®
Lieferant: VWR Chemicals
|
Gefahr
|
23470.293EA
57.2
EUR
23470.293
23470.327
23470.442
23470.930
N,N-Dimethylformamid ≥99,5%, GPR RECTAPUR®
N,N-Dimethylformamid
Formel:
HCON(CH₃)₂ Siedepunkt: 153 °C (1013 hPa) Schmelzpunkt: –60,5 °C Dichte: 0,949 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003284 CAS-Nummer: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3,III REACH: 01-2119475605-32 Merck Index: 13,03269 |
|
Spezifikation Testergebnisse
Assay | Min. 99.5 % |
Appearance | Clear colourless liquid |
IR Spectrum | Passes test |
Acidity | Max. 0.00025 meq/g |
Alkalinity | Max. 0.003 meq/g |
Boiling point | 152 - 155 °C |
Density (20/4) | 0.940 - 0.950 |
Evaporation residue | Max. 100 ppm |
Water | Max. 0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...