Acridinorange Base ≥75% (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Brilliant Acridine Orange E, 3,6-Bis(dimethylamino)acridine, Acridine Orange base, Solvent Orange 15, 3,6-Bis(dimethylamino)acridine N,N,N',N'-Tetramethyl-3,6-acridinediamine, Acridine orange free base, Acridine Orange Stain

M-Clarity™ Qualitätsstufe = MQ200

235474-25G 235474-5G
235474-25GEA 273 EUR
235474-25G 235474-5G
Acridinorange Base ≥75% (Farbstoffgehalt), Sigma-Aldrich®
Acridinorange

Acridine Orange base (3,6-Bis(dimethylamino)acridine) is a cell-permeable fluorescent dye that binds to nucleic acids. It emits green fluorescence when bound to dsDNA and red fluorescence when bound to ssDNA or RNA. It is also known to be a positive solvatochromic dye. Acridine orange has also been used as a lysosomal dye. It is widely used to detect the presence of homeopathic potencies.

Formel: C₁₇H₁₉N₃
Molecular Weight: 265.35 g/mol
Schmelzpunkt: 165 °C (Zers., Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005029
CAS-Nummer: 494-38-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=888823beccf53209a405a0fa2635aa17c7031fa8&vid=4d03cc4a6dbc866c2c25327340e9b2728c334a9a&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wavelength 227 - 233 nm
Extinction Coefficient ≥14000
Carbon ≥56.2 %
Nitrogen ≥11.6 %
Purity (HPLC) ≥75 %
Solubility (Turbidity) Clear
Solubility (Color); Orange to Very Dark Red-Brown, c = 1mg/ml, 1N HCl Conforms to Requirements


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR