Kieselgel, Davisil® Grade 643 pore size 60 Å, 35-60 mesh particle size, hochrein, Supelco®

Lieferant: Merck

236802-100G
236802-100GEA 93.1 EUR
236802-100G
Kieselgel, Davisil® Grade 643 pore size 60 Å, 35-60 mesh particle size, hochrein, Supelco®
Silica gel
Formel: SiO₂
Siedepunkt: 2230 °C
Schmelzpunkt: > 1600 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011232
CAS-Nummer: 63231-67-4
EINECS: 231-545-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96fdbe3af011ec5fc1e18cbd73cbd9daa2957903&vid=ba488c2a4e2cbd43e2df1646d53cfdb1d9af9f09&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR