Kieselgel, Davisil® Grade 646 pore size 150 Å, 200-425 mesh, hochrein, Supelco®

Lieferant: Merck

236810-100G 236810-1KG
236810-100GEA 106 EUR
236810-100G 236810-1KG
Kieselgel, Davisil® Grade 646 pore size 150 Å, 200-425 mesh, hochrein, Supelco®
Silica gel
Formel: SiO₂
Siedepunkt: 2230 °C
Schmelzpunkt: > 1600 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011232
CAS-Nummer: 63231-67-4
EINECS: 231-545-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98fa2cfba5aac75b8ecb042746613f3bd47bb9c8&vid=1af319f6f202ec01ce96255ad4f2bf1a80d0c9b9&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR