Kieselgel, Davisil® Grade 646 pore size 150 Å, 200-425 mesh, hochrein, Supelco®
Lieferant: Merck
236810-100G
236810-1KG
236810-100GEA
106
EUR
236810-100G
236810-1KG
Kieselgel, Davisil® Grade 646 pore size 150 Å, 200-425 mesh, hochrein, Supelco®
Silica gel
Formel:
SiO₂ Siedepunkt: 2230 °C Schmelzpunkt: > 1600 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011232 CAS-Nummer: 63231-67-4 EINECS: 231-545-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...