Kieselgel, Davisil® Grade 62 pore size 150 Å, 35-60 mesh, hochrein, Supelco®

Lieferant: Merck

236845-100G 236845-1KG
236845-100GEA 100 EUR
236845-100G 236845-1KG
Kieselgel, Davisil® Grade 62 pore size 150 Å, 35-60 mesh, hochrein, Supelco®
Silica gel
Formel: SiO₂
Siedepunkt: 2230 °C
Schmelzpunkt: > 1600 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011232
CAS-Nummer: 63231-67-4
EINECS: 231-545-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc93c274583970dc8baa52abfe2cf9ddb1f7c5d9&vid=9bbeb82d81a0a3c04adfd63ae4db4fd0eb202793&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR