Lithiumchlorid ≥99,0% (durch argentometrische Titration), körnig, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
|
Achtung
|
2370-07EA
0
EUR
2370-07
Lithiumchlorid ≥99,0% (durch argentometrische Titration), körnig, BAKER ANALYZED® ACS, J.T.Baker®
Lithium chloride
Formel:
LiCl Molecular Weight: 42,39 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011078 CAS-Nummer: 7447-41-8 |
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use |
|
ACS - Assay (LiCl) (by Ag titrn) | ≥ 99.0 % |
ACS - Insoluble Matter | ≤ 0.01 % |
ACS - Loss on Drying | ≤ 1.0 % |
ACS - Titrable Base (meq/g) | ≤ 0.008 |
ACS - Sulfate (SO₄) | ≤ 0.01 % |
ACS - Barium (Ba) | ≤ 0.002 % |
ACS - Heavy Metals (as Pb) | ≤ 0.002 % |
ACS - Iron (Fe) | ≤ 0.001 % |
ACS - Calcium (Ca) | ≤ 0.01 % |
ACS - Potassium (K) | ≤ 0.01 % |
ACS - Sodium (Na) | ≤ 0.20 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...