Insulation stripper
Lieferant: KNIPEX WERKE
1106160
238-5410EA 49.4 EUR
238-5410
Insulation stripper
Zangen
Chrome plated, forged, oil-hardened insulation stripper.
  • For single, multiple and fine stranded conductors with plastic or rubber insulation max. Ø 5,0 mm or respectively 10 mm² cable cross section
  • Easy adjustment to the required diameter of solid or stranded wire with knurled screw and lock nut
  • Handles insulated with multi-component grips
Standards: DIN EN 60900, IEC 60900. VDE-tested