The plier wrench combines the jobs of a gripping plier and a wrench in a single tool with flexible adjustment of all widths up to the specified maximum size. Ideal for for gripping, holding, pressing and bending workpieces.
Adjustable tightening tool
Parallel jaws give a more solid grip
Zero backlash jaw pressure prevents damage to edges
Easily adjusted at the touch of a button on the workpiece
Smooth jaws for damage free installation of plated fittings