Creatine phosphate disodium salt, Millipore®

Lieferant: Merck Millipore Calbiochem

2380-25 2380-5
2380-25EA 358 EUR
2380-25 2380-5
Creatine phosphate disodium salt, Millipore®
Creatinphosphat Dinatriumsalz Hydrat
Formel: C₄H₈N₃Na₂O₅P·H₂O
MDL: MFCD00150191
CAS-Nummer: 19333-65-4
EINECS: 213-074-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR