Creatine phosphate disodium salt, Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
Phosphocreatine disodium hydrate, Sodium 2-(1-methyl-3-phosphonatoguanidino)acetate hydrate(2:1:x), N-(Imino[phosphonoamino]methyl)-N-methylglycine hydrate, Phosphocreatine disodium salt hydrate
2380-25
2380-5
2380-25EA
358
EUR
2380-25
2380-5
Creatine phosphate disodium salt, Millipore®
Creatinphosphat Dinatriumsalz Hydrat
Formel:
C₄H₈N₃Na₂O₅P·H₂O Molecular Weight: 273.09 g/mol |
MDL:
MFCD00150191 CAS-Nummer: 19333-65-4 EINECS: 213-074-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...