Diethylether ≥99,7% stabilisiert, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens

Lieferant: VWR Chemicals
Gefahr


Stabilisierung: Stabilisiert mit BHT/Ionol/2,6-Di-tert-butyl-4-methylphenol 4-10 ppm

23811.292EA 43.2 EUR
23811.292 23811.361 23811.326 23811.463 23811.550
Diethylether ≥99,7% stabilisiert, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens
Diethylether
Formel: (CH₃CH₂)₂O
Siedepunkt: 35 °C (1013 hPa)
Schmelzpunkt: –123 °C
Dichte: 0,7135 g/cm³ (20 °C)
Flammpunkt: –40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011646
CAS-Nummer: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay Min. 99.7 %
Assay (Ionol/BHT) 4.0 - 10.0 ppm
Aldehydes Not detected by GC
Insolubility in water Passes test
IR Spectrum Passes test
Acidity Max. 0.0001 meq/g
Boiling point 34 - 35 °C
Colouration Max. 10 APHA
Relative density 0.714 - 0.715
Substances coloured by H2SO4 Max. 10 APHA
Acetone Max. 10 ppm
Aldehydes + ketones (as HCHO) Max. 5 ppm
Carbonyl compounds (as HCHO) Max. 10 ppm
Ethanol Max. 0.05 %
Evaporation residue Max. 5 ppm
Methanol Max. 0.02 %
Peroxides (as H2O2) Max. 0.15 ppm
Water Max. 300 ppm
Al (Aluminium) Max. 0.5 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.05 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 0.1 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.1 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.1 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR