Diethylether ≥99,7% stabilisiert, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens
Lieferant: VWR Chemicals
|
Gefahr
|
Stabilisierung:
Stabilisiert mit BHT/Ionol/2,6-Di-tert-butyl-4-methylphenol 4-10 ppm
23811.292EA
72
EUR
23811.292
23811.361
23811.326
23811.463
23811.550
Diethylether ≥99,7% stabilisiert, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens
Diethylether
Formel:
(CH₃CH₂)₂O Siedepunkt: 35 °C (1013 hPa) Schmelzpunkt: –123 °C Dichte: 0,7135 g/cm³ (20 °C) Flammpunkt: –40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011646 CAS-Nummer: 60-29-7 EINECS: 200-467-2 UN: 1155 ADR: 3,I REACH: 01-2119535785-29 Merck Index: 12,03852 |
|
Spezifikation Testergebnisse
Assay | Min. 99.7 % |
Assay (Ionol/BHT) | 4.0 - 10.0 ppm |
Aldehydes | Not detected by GC |
Insolubility in water | Passes test |
IR Spectrum | Passes test |
Acidity | Max. 0.0001 meq/g |
Boiling point | 34 - 35 °C |
Colouration | Max. 10 APHA |
Relative density | 0.714 - 0.715 |
Substances coloured by H2SO4 | Max. 10 APHA |
Acetone | Max. 10 ppm |
Aldehydes + ketones (as HCHO) | Max. 5 ppm |
Carbonyl compounds (as HCHO) | Max. 10 ppm |
Ethanol | Max. 0.05 % |
Evaporation residue | Max. 5 ppm |
Methanol | Max. 0.02 % |
Peroxides (as H2O2) | Max. 0.15 ppm |
Water | Max. 300 ppm |
Al (Aluminium) | Max. 0.5 ppm |
Ba (Barium) | Max. 0.1 ppm |
Ca (Calcium) | Max. 0.5 ppm |
Cd (Cadmium) | Max. 0.05 ppm |
Co (Cobalt) | Max. 0.02 ppm |
Cr (Chromium) | Max. 0.02 ppm |
Cu (Copper) | Max. 0.02 ppm |
Fe (Iron) | Max. 0.1 ppm |
K (Potassium) | Max. 0.05 ppm |
Mg (Magnesium) | Max. 0.1 ppm |
Mn (Manganese) | Max. 0.02 ppm |
Na (Sodium) | Max. 0.1 ppm |
Ni (Nickel) | Max. 0.02 ppm |
Pb (Lead) | Max. 0.1 ppm |
Sn (Tin) | Max. 0.1 ppm |
Sr (Strontium) | Max. 0.02 ppm |
Zn (Zinc) | Max. 0.1 ppm |
Conforms to ACS | Passes test |
Conforms to Reag. Ph.Eur. | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...