Diethylether stabilisiert Ph. Eur.

Lieferant: VWR Chemicals
Gefahr


Stabilisierung: Stabilisiert mit BHT/Ionol/2,6-Di-tert-butyl-4-methylphenol

23819.298EA 75.2 EUR
23819.298 23819.367 23819.460
Diethylether stabilisiert Ph. Eur.
Diethylether

Appearance: Clear, colourless, volatile, very mobile liquid.


Bitte beachten Not for use as Active Pharmaceutical Ingredient (API)

Formel: (CH₃CH₂)₂O
Siedepunkt: 35 °C (1013 hPa)
Schmelzpunkt: –123 °C
Dichte: 0,7135 g/cm³ (20 °C)
Flammpunkt: –40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011646
CAS-Nummer: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (Ionol/BHT) Passes test
Appearance Conforms (see CoA/CoS)
Acidity Passes test
Relative density 0.714 - 0.716
Distillation range 34.0 - 35.0 °C
Acetone and aldehydes Passes test
Peroxides Passes test
Non-volatile matter ≤ 20 mg/l
Unspecified impurities ≤ 0.05 %
Total impurities ≤ 0.1 %
Reporting threshold ≤ 0.03 %
Water ≤ 2 g/l
Residual solvents Passes test
As (Arsenic) ≤ 0.1 ppm
Cd (Cadmium) ≤ 0.1 ppm
Co (Cobalt) ≤ 0.1 ppm
Hg (Mercury) ≤ 0.1 ppm
Ni (Nickel) ≤ 0.1 ppm
Pb (Lead) ≤ 0.1 ppm
V (Vanadium) ≤ 0.1 ppm
Conforms to Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR