Natriumsulfat, anhydrous 99,0, powder ACS, Fluka™

Lieferant: Honeywell Chemicals

Synonyme: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt

238597-2.5KG 238597-500G 238597-1KG
238597-2.5KGEA 117.08 EUR
238597-2.5KG 238597-500G 238597-1KG
Natriumsulfat, anhydrous 99,0, powder ACS, Fluka™
Natriumsulfat
Formel: Na₂SO₄
Molecular Weight: 142.04 g/mol
Siedepunkt: 1429
Schmelzpunkt: 884 °C
Dichte: 2.68 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003504
CAS-Nummer: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Identity complying
Assay Min. 99.0 %
pH (5 %, 25 °C) 5.2 - 9.2
Loss on ignition (800°C) Max. 0.5 %
Water insoluble matter Max. 0.01 %
Calcium (Ca) Max. 0.01 %
Iron (Fe) Max. 0.001 %
Potassium (K) Max. 0.01 %
Magnesium (Mg) Max. 0.005 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 0.001 %
Phosphate (PO₄) Max. 0.001 %
Total N Max. 5 ppm
pH Range 5.2 - 9.2
Form powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR